COMH11E51E16 Combi-Blocks, Inc.

4,6-Difluoro-1-methyl-1h-indazole

  • MDL number MFCD16658676
  • Cas Number : 1185767-08-1
  • Molecular Weight : 168.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-0916

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-0916
Availability details for
Quantity
  • Availability
  • Add to Cart