COMH11E51E1B Combi-Blocks, Inc.

2-(2,6-Dichlorophenoxy)propanoic acid

  • MDL number MFCD03944281
  • Cas Number : 25140-90-3
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5451
Availability details for
Quantity
  • Availability
  • Add to Cart