COMH11E51E1D Combi-Blocks, Inc.

Methyl 3-(bromomethyl)-5-fluorobenzoate

  • MDL number MFCD22988208
  • Cas Number : 816449-87-3
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3485
Availability details for
Quantity
  • Availability
  • Add to Cart