COMH11E51E1E Combi-Blocks, Inc.

9-(2-Chloro-ethyl)-9H-purin-6-ylamine

  • MDL number MFCD00823497
  • Cas Number : 19255-48-2
  • Molecular Weight : 197.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5732
Availability details for
Quantity
  • Availability
  • Add to Cart