COMH11E51E22 Combi-Blocks, Inc.

(E)-Ethyl 3-(1-methyl-1H-pyrazol-4-yl)acrylate

  • MDL number MFCD23715484
  • Cas Number : 192661-36-2
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8808
Availability details for
Quantity
  • Availability
  • Add to Cart