COMH11E51E28 Combi-Blocks, Inc.

Ethyl 2-amino-4,4,4-trifluoro-3-oxobutanoate hydrochloride

  • MDL number MFCD28969400
  • Cas Number : 945382-02-5
  • Molecular Weight : 235.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6539
Availability details for
Quantity
  • Availability
  • Add to Cart