COMH11E51E2B Combi-Blocks, Inc.

{[4-(benzyloxy)phenyl]formamido}acetic acid

  • MDL number MFCD01912116
  • Cas Number : 171806-97-6
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6826
Availability details for
Quantity
  • Availability
  • Add to Cart