COMH11E51E2F Combi-Blocks, Inc.

2-N,2-N-diethylpyridine-2,4-diamine

  • MDL number MFCD13190682
  • Cas Number : 90768-40-4
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6854
Availability details for
Quantity
  • Availability
  • Add to Cart