COMH11E51E31 Combi-Blocks, Inc.

2-N-phenylpyridine-2,4-diamine

  • MDL number MFCD16084053
  • Cas Number : 25194-51-8
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6856
Availability details for
Quantity
  • Availability
  • Add to Cart