COMH11E51E34 Combi-Blocks, Inc.

1-(4-bromo-2-fluorophenyl)-4-methylpiperazine

  • MDL number MFCD20919836
  • Cas Number : 1352207-52-3
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6895
Availability details for
Quantity
  • Availability
  • Add to Cart