COMH11E51E38 Combi-Blocks, Inc.

(4-Chloro-2-fluoro-6-methoxyphenyl)boronic acid

  • MDL number MFCD30329340
  • Cas Number : 1628684-10-5
  • Molecular Weight : 204.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6928

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6928
Availability details for
Quantity
  • Availability
  • Add to Cart