COMH11E51E39 Combi-Blocks, Inc.

2-(3-iodopropyl)isoindole-1,3-dione

  • MDL number MFCD00458300
  • Cas Number : 5457-29-4
  • Molecular Weight : 315.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6929
Availability details for
Quantity
  • Availability
  • Add to Cart