COMH11E51E3A Combi-Blocks, Inc.

Ethyl 7-bromo-1-oxo-3,4-dihydro-2H-naphthalene-2-carboxylate

  • MDL number MFCD24539298
  • Cas Number : 914262-72-9
  • Molecular Weight : 297.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6939

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6939
Availability details for
Quantity
  • Availability
  • Add to Cart