COMH11E51E3D Combi-Blocks, Inc.

N-[(1Z)-2-pentylcyclopentylidene]hydroxylamine

  • MDL number MFCD02151982
  • Cas Number : 1419029-15-4
  • Molecular Weight : 169.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6942
Availability details for
Quantity
  • Availability
  • Add to Cart