COMH11E51E60 Combi-Blocks, Inc.

2-Phenyl-ethenesulfonic acid amide

  • MDL number MFCD00118353
  • Cas Number : 4363-41-1
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8644

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8644
Availability details for
Quantity
  • Availability
  • Add to Cart