COMH2F649CC8 Combi-Blocks, Inc.

4-(4-BOC-Piperidin-3-yloxy)phenylboronic acid

  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1573

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1573
Availability details for
Quantity
  • Availability
  • Add to Cart