COMH2F649CCE Combi-Blocks, Inc.

N-(3,5-Dichloropyridin-4-yl)-3,4-difluorobenzamide

  • MDL number MFCD32065681
  • Cas Number : 2365418-74-0
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6036
Availability details for
Quantity
  • Availability
  • Add to Cart