COMH2F649CD7 Combi-Blocks, Inc.

4-Bromo-N-methyl-1,8-naphthalimide

  • MDL number MFCD00446021
  • Cas Number : 4116-90-9
  • Molecular Weight : 290.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6049

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6049
Availability details for
Quantity
  • Availability
  • Add to Cart