COMH2F649D16 Combi-Blocks, Inc.

4-Bromo-3-(trifluoromethyl)-1-(triphenylmethyl)pyrazole

  • MDL number MFCD13189512
  • Cas Number : 474706-50-8
  • Molecular Weight : 457.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0583
Availability details for
Quantity
  • Availability
  • Add to Cart