COMH2F649D38 Combi-Blocks, Inc.

1-{[2-(Benzyloxy)phenyl]methyl}piperazine

  • MDL number MFCD05189153
  • Cas Number : 523980-11-2
  • Molecular Weight : 282.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[2-(benzyloxy)benzyl]piperazine
Inchi Code 1S/C18H22N2O/c1-2-6-16(7-3-1)15-21-18-9-5-4-8-17(18)14-20-12-10-19-11-13-20/h1-9,19H,10-15H2
Inchi Key CRNNQUKZDCTDKW-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2326
Availability details for
Quantity
  • Availability
  • Add to Cart