COMH2F649D3B Combi-Blocks, Inc.

4-(2-Nitrophenoxy)piperidine

  • MDL number MFCD16076675
  • Cas Number : 1073055-66-9
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2336
Availability details for
Quantity
  • Availability
  • Add to Cart