COMH2F649D3C Combi-Blocks, Inc.

4-((2-Nitrophenoxy)methyl)piperidine

  • MDL number MFCD12407024
  • Cas Number : 63608-16-2
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2337
Availability details for
Quantity
  • Availability
  • Add to Cart