COMH2F649D42 Combi-Blocks, Inc.

4-(4-Fluoro-3-(trifluoromethyl)phenoxy)piperidine

  • MDL number MFCD18632624
  • Cas Number : 1188375-44-1
  • Molecular Weight : 263.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2343

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2343
Availability details for
Quantity
  • Availability
  • Add to Cart