COMH2F649D43 Combi-Blocks, Inc.

4-((4-Fluoro-3-(trifluoromethyl)phenoxy)methyl)piperidine

  • MDL number MFCD32065694
  • Cas Number : 1974995-21-5
  • Molecular Weight : 277.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2344
Availability details for
Quantity
  • Availability
  • Add to Cart