COMH2F649D7A Combi-Blocks, Inc.

(5-Ethyl-2-methoxyphenyl)methanol

  • MDL number MFCD21909890
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4623
Availability details for
Quantity
  • Availability
  • Add to Cart