COMH2F649D7B Combi-Blocks, Inc.

2-(Chloromethyl)-4-ethyl-1-methoxybenzene

  • MDL number MFCD20624321
  • Cas Number : 85944-01-0
  • Molecular Weight : 184.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4624

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4624
Availability details for
Quantity
  • Availability
  • Add to Cart