COMH2F649D86 Combi-Blocks, Inc.

2-(2-methoxy-4-methylphenyl)acetonitrile

  • MDL number MFCD18398344
  • Cas Number : 1261796-01-3
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4638
Availability details for
Quantity
  • Availability
  • Add to Cart