COMH2F649D88 Combi-Blocks, Inc.

5-chloro-2-methoxy-4-methylbenzaldehyde

  • MDL number MFCD06246304
  • Cas Number : 82128-70-9
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-methoxy-4-methylbenzaldehyde
Inchi Code 1S/C9H9ClO2/c1-6-3-9(12-2)7(5-11)4-8(6)10/h3-5H,1-2H3
Inchi Key QIFCXAISIRTSGA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4640
Availability details for
Quantity
  • Availability
  • Add to Cart