COMH2F649D8B Combi-Blocks, Inc.

1,2,3,4-Tetrahydroquinoline-7-carbonitrile

  • MDL number MFCD08544273
  • Cas Number : 939758-76-6
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4643
Availability details for
Quantity
  • Availability
  • Add to Cart