COMH2F649D8E Combi-Blocks, Inc.

4-(2-aminoethoxy)benzoic acid

  • MDL number MFCD02018963
  • Cas Number : 52660-66-9
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4648

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4648
Availability details for
Quantity
  • Availability
  • Add to Cart