COMH2F649D8F Combi-Blocks, Inc.

Methyl 3-formamido-4-methoxybenzoate

  • MDL number MFCD32065710
  • Cas Number : 2279976-13-3
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4651

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4651
Availability details for
Quantity
  • Availability
  • Add to Cart