COMH2F649D94 Combi-Blocks, Inc.

7-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid

  • MDL number MFCD32065763
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1562

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1562
Availability details for
Quantity
  • Availability
  • Add to Cart