COMH2F649D97 Combi-Blocks, Inc.

4-Fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-dicarboxylic acid

  • Molecular Weight : 310.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1572
Availability details for
Quantity
  • Availability
  • Add to Cart