COMH2F649DB1 Combi-Blocks, Inc.

Methyl 5-hexynoate

  • MDL number MFCD00671366
  • Cas Number : 77758-51-1
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4115
Availability details for
Quantity
  • Availability
  • Add to Cart