COMH2F649DD2 Combi-Blocks, Inc.

N-Propyl chlorothioformate

  • MDL number MFCD00018807
  • Cas Number : 13889-92-4
  • Molecular Weight : 138.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1716

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1716
Availability details for
Quantity
  • Availability
  • Add to Cart