COMH2F649DD3 Combi-Blocks, Inc.

N1-Boc-diethylenetriamine

  • MDL number MFCD06797591
  • Cas Number : 193206-49-4
  • Molecular Weight : 203.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1987

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1987
Availability details for
Quantity
  • Availability
  • Add to Cart