COMH303775DA Combi-Blocks, Inc.

2-Ethoxy-3-(trifluoromethoxy)aniline

  • MDL number MFCD32065661
  • Cas Number : 2168783-23-9
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6358

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6358
Availability details for
Quantity
  • Availability
  • Add to Cart