COMH303775DE Combi-Blocks, Inc.

2-Carboxy-7-methylimidazo[1,2-a]pyridine-6-boronic acid

  • MDL number MFCD32065665
  • Cas Number : 2377609-21-5
  • Molecular Weight : 220.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1102
Availability details for
Quantity
  • Availability
  • Add to Cart