COMH303775E3 Combi-Blocks, Inc.

7-bromo-4,5-difluoro-1,3-dihydroindol-2-one

  • MDL number MFCD31537024
  • Cas Number : 2090777-89-0
  • Molecular Weight : 248.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0590
Availability details for
Quantity
  • Availability
  • Add to Cart