COMH303775E6 Combi-Blocks, Inc.

Methyl 2-(4-bromo-2-nitrophenyl)-2-methylpropanoate

  • MDL number MFCD29053109
  • Molecular Weight : 302.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0593
Availability details for
Quantity
  • Availability
  • Add to Cart