COMH303775EF Combi-Blocks, Inc.

[2-(1,3-Benzothiazol-2-yloxy)ethyl](methyl)amine

  • MDL number MFCD16713844
  • Molecular Weight : 208.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[2-(1,3-benzothiazol-2-yloxy)ethyl]-N-methylamine
Inchi Code 1S/C10H12N2OS/c1-11-6-7-13-10-12-8-4-2-3-5-9(8)14-10/h2-5,11H,6-7H2,1H3
Inchi Key NLGOWGBNJUCCMU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2380
Availability details for
Quantity
  • Availability
  • Add to Cart