COMH303775F1 Combi-Blocks, Inc.

2-(Piperidin-3-yloxy)-1,3-benzothiazole

  • MDL number MFCD16736588
  • Cas Number : 1185540-94-6
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2384
Availability details for
Quantity
  • Availability
  • Add to Cart