COMH30377611 Combi-Blocks, Inc.

5,6-Dichloroquinazolin-2-amine

  • MDL number MFCD32065711
  • Cas Number : 2365420-03-5
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4656
Availability details for
Quantity
  • Availability
  • Add to Cart