COMH30377613 Combi-Blocks, Inc.

(5-Chloro-2-fluoro-4-methylphenyl)methanol

  • MDL number MFCD27948131
  • Cas Number : 252004-44-7
  • Molecular Weight : 174.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4658
Availability details for
Quantity
  • Availability
  • Add to Cart