COMH30377616 Combi-Blocks, Inc.

6-Bromo-3,4-dihydro-1H-isoquinoline-2-carbaldehyde

  • MDL number MFCD32065712
  • Cas Number : 2365419-41-4
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4666
Availability details for
Quantity
  • Availability
  • Add to Cart