COMH30377620 Combi-Blocks, Inc.

3-[(tert-butyldimethylsilyl)oxy]butan-2-one

  • MDL number MFCD32065713
  • Cas Number : 108269-33-6
  • Molecular Weight : 202.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4680
Availability details for
Quantity
  • Availability
  • Add to Cart