COMH30377621 Combi-Blocks, Inc.

Ethyl (2E)-4,4-dimethoxy-3-methylpent-2-enoate

  • MDL number MFCD32065714
  • Cas Number : 98315-32-3
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4681
Availability details for
Quantity
  • Availability
  • Add to Cart