COMH30377624 Combi-Blocks, Inc.

2-(4-Methoxynaphthalen-1-yl)acetonitrile

  • MDL number MFCD18412854
  • Cas Number : 177598-50-4
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4685

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4685
Availability details for
Quantity
  • Availability
  • Add to Cart