COMH30377627 Combi-Blocks, Inc.

3-Bromo-2,6-difluoro-5-methylphenol

  • MDL number MFCD32065715
  • Cas Number : 1780848-69-2
  • Molecular Weight : 223.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4689

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4689
Availability details for
Quantity
  • Availability
  • Add to Cart