COMH30377632 Combi-Blocks, Inc.

Ethyl 5-bromo-2,3-dihydro-1-benzofuran-2-carboxylate

  • MDL number MFCD15523670
  • Cas Number : 194163-36-5
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4704

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4704
Availability details for
Quantity
  • Availability
  • Add to Cart